• USA Home
  • Z266760 - Stirring shaft 316 SS
EMAIL THIS PAGE TO A FRIEND
Z266760 Aldrich
Stirring shaft 316 SS
shaft diam. × L 3/8 in. × 24 in.
Purchase
Properties
Related Categories Barnant, Barnant Stirrers, Labware, Manufacturer Browser, Overhead - SS Stirrers,
mfr. no.   Barnant
shaft diam. × L   3/8 in. × 24 in.
Description
General description
The 3/8 in. diam. shafts are machined on one end to also accept 5/16 in. bore diam. propellers.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?