• USA Home
  • Z267007 - Balances, Sartorius high-capacity top-loading Model BP12000S
EMAIL THIS PAGE TO A FRIEND
Z267007 Aldrich
Balances, Sartorius high-capacity top-loading Model BP12000S
Model BP12000S, AC/DC input 220 V AC
Purchase
Properties
Related Categories Balances and Accessories, Labware, Sartorius Balances More...
mfr. no.   Model BP12000S
availability   not available in USA and Canada
measuring range   12000 g weighing capacity, resolution: 0.1 g
size   417 mm × 307 mm
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?