• USA Home
  • Z268380 - Pall glass-fiber filters
EMAIL THIS PAGE TO A FRIEND
Z268380 Aldrich
Pall glass-fiber filters
diam. 90 mm
Purchase
Properties
Related Categories Filtration Supplies, Glass Fiber Filters, Labware, Manufacturer Browser, Pall Gelman,
material   borosilicate glass membrane
  circles
packaging   pkg of 100 ea
mfr. no.   Gelman 61664
diam.   90 mm
pore size   1 μm
Description
General description
Glass-fiber filters are ideal for prefiltration, sample cleanup, and many analytical applications. Binder-free borosilicate glass filters offer a high degree of wet strength.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?