• USA Home
  • Z268399 - Pall glass-fiber filters
EMAIL THIS PAGE TO A FRIEND
Z268399 Aldrich
Pall glass-fiber filters
TCLP, diam. 47 mm
Purchase
Properties
Related Categories Filtration Supplies, Glass Fiber Filters, Labware, Manufacturer Browser, Pall Gelman,
material   borosilicate glass membrane
  circles
packaging   pkg of 50 ea
mfr. no.   Gelman 66258
diam.   47 mm
pore size   0.7 μm
suitability   suitable for (for EPA Method 1311, Toxicity Characteristics Leaching Procedure)
Description
General description
Glass-fiber filters are ideal for prefiltration, sample cleanup, and many analytical applications. Binder-free borosilicate glass filters offer a high degree of wet strength.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?