• USA Home
  • Z268410 - Pall reusable syringe filter holders
EMAIL THIS PAGE TO A FRIEND
Z268410 Aldrich
Pall reusable syringe filter holders
diam. 13 mm
  •  eCl@ss 32110206
Purchase
Properties
Related Categories Filtration Supplies, Labware, Manufacturer Browser, Membrane Holders and Assemblies, Pall Gelman,
material   polyacetal (resin construction)
  polypropylene housing (housing)
sterility   non-sterile
mfr. no.   Gelman 4317
packaging   pkg of 5 ea
diam.   13 mm
pore size   (N/A)
fitting   inlet and outlet Luer
Description
General description
Reusable plastic filter holders provide an affordable alternative to disposable syringe filters. Autoclavable.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?