• USA Home
  • Z268690 - Scienceware® Safe-Lab PTFE stopper
EMAIL THIS PAGE TO A FRIEND
Z268690 Aldrich
Scienceware® Safe-Lab PTFE stopper
Joint: ST/NS 19/22, solid
Synonym: lab stopper, lab stoppers, stoppers
Purchase
Properties
Related Categories Bel-Art, Bel-Art Stoppers, Labware, Manufacturer Browser, Plastic Stoppers,
Joint   Joint: ST/NS19/22
packaging   pkg of 3 ea
mfr. no.   Bel-Art
Description
General description
PTFE stoppers with PP extracting nut for easy removal from flask. For use with boiling and distilling flasks with ground joints.
Legal Information
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?