• USA Home
  • Z268836 - Scienceware® Safe-Lab vacuum adapter
EMAIL THIS PAGE TO A FRIEND
Z268836 Aldrich
Scienceware® Safe-Lab vacuum adapter
Joint: ST/NS 14/20
Purchase
Description
General description
PTFE body with Viton® O-ring. Hose barb connection for vacuum work or inlet/outlet of gases.
Legal Information
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?