• USA Home
  • Z268909 - Safe-Lab Therm-O-Vac® port adapter
EMAIL THIS PAGE TO A FRIEND
Z268909 Aldrich
Safe-Lab Therm-O-Vac® port adapter
opening size 6 mm
Purchase
Description
General description
Threaded PTFE adapter body with side hose barb for vacuum applications. For use with the PTFE Adapt-A-Port® joint adapters.
Legal Information
Safe-Lab is a trademark of SP Scienceware, a division of SP
Therm-O-Vac is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?