• USA Home
  • Z268976 - Scienceware® Poly-Cone adapters
EMAIL THIS PAGE TO A FRIEND
Z268976 Aldrich
Scienceware® Poly-Cone adapters
Purchase
Properties
Related Categories Adapters, Bel-Art, Bel-Art Adapters, Gas Equipment Adapters, Labware,
mfr. no.   Bel-Art
Description
General description
Universal PP adapter forms a continuous taper from top to bottom, both inside and out. May be cut with a knife or saw into a variety of adapters. Allows connection of two equal or unequal joints. Overall length: 27 cm and the diameter ranges from: 35 to 5 mm with a standard taper.
Legal Information
Poly-Cone is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?