Z269271 Aldrich

Pall membrane filters

HT Tuffryn membrane, diam. 25 mm, pore size 0.2 μm

DOWNLOAD MSDS (PDF)

Properties

Related Categories Filtration Supplies, Labware, Manufacturer Browser, Membrane Filters, Pall Gelman,
material   HT Tuffryn membrane
mfr. no.   Gelman 66197
packaging   pkg of 100 ea
diam.   25 mm
pore size   0.2 μm

Description

General description

Offer a wide variety of membranes that meet almost any filtration need.
Supor® polyethersulfone, for biological and pharmaceutical solutions, fast flow, low extractables, autoclavable.
HT Tuffryn® polysulfone, for media and air samples, low-protein-binding, autoclavable.
GN Metricel® mixed cellulose esters, for air samples and microbiological analysis, meets US EPA and ASTM specifications, autoclavable.
Nylaflo nylon, for general-purpose applications, HPLC, good chemical resistance, low extractables.
PTFE PTFE on PP support, for aggressive organics, HPLC and air/gases, hydrophobic, chemically inert.

Legal Information

Metricel is a registered trademark of Pall Corporation

Nylaflo is a trademark of Pall Corporation

Supor is a registered trademark of Pall Corporation

Tuffryn is a registered trademark of Pall Corporation

Price and Availability

Customers Also Viewed

Acrodisc<SUP>®</SUP> syringe filters

Nylon membrane, diam. 13 mm, pore size 0.2 μm

Acrodisc<SUP>®</SUP> syringe filters

HT Tuffryn® membrane, diam. 25 mm, porosity 0.45 μm

Pall membrane filters

HT Tuffryn membrane, diam. 25 mm, pore size 0.45 μm

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?