• USA Home
  • Z269581 - Safety pipette fillers
EMAIL THIS PAGE TO A FRIEND
Z269581 Aldrich
Safety pipette fillers
blue
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Pipettes, Pipettors and Tips, Pipettors, Sigma Pipettors More...
material   natural rubber
color   blue
Description
General description
Has two operating valves to fill and dispense and a third removable automatic valve at the top for evacuation and easy inside cleaning. Natural rubber.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?