• USA Home
  • Z269662 - Biodegradable Hydrogels for Drug Delivery
EMAIL THIS PAGE TO A FRIEND
Z269662 Aldrich
Biodegradable Hydrogels for Drug Delivery
  •  ISBN-10 1566760046
  •  ISBN-13 978-156676004-1
Purchase
Properties
Related Categories Biotechnology Books, Books and Software, Labware, Pharmaceutical and Medicinal Chemistry Books
publication info   K. Park, et al., Technomic Publishing Co., 1993, 252 pp., hard cover
mfr. no.   CRC Press
Description
General description
This book provides an in-depth investigation of recent advances in biodegradable hydrogels. Based on extensive research and experience, the book is the first to deal exclusively with this important area in drug delivery. A valuable guide for all, the book begins with a detailed introduction to current biodegradable hydrogel technology. Following chapters present advanced R&D, delivery systems, and more. It covers the mechanisms of biodegradation, types of biodegradable hydrogels, chemical and physical gels, chemical and enzymatic degradation, and examples of biodegradable drug delivery systems.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?