EMAIL THIS PAGE TO A FRIEND
Z269786 Aldrich
Thermistor sensor
L 5 in.
Purchase
Properties
material   glass
availability   not available in EU
L   5 in.
Description
General description
Use with digital cold plates and cold plate stirrers for automatic temperature monitoring and control.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?