• USA Home
  • Z269921 - Plas-Labs Nitrogen dry-box glove box
EMAIL THIS PAGE TO A FRIEND
Z269921 Aldrich
Plas-Labs Nitrogen dry-box glove box
AC input 110 V
Purchase
Properties
grade   CE Compliant
mfr. no.   Plas-Labs 850-NB/SP800-SHF-I
H × W × D   26 in. × 41 in. × 26 in., inside dimensions
  38 in. × 55 in. × 35 in., outside dimensions
weight   250 lb
Description
General description
The Plas-Labs nitrogen single-station glove box is a completely enclosed chamber which is ideal for working in a preferred atmosphere. Inert gases like Argon, Nitrogen, and Plasma are easily maintained. It is compact, portable, and lightweight. The design makes is easily moveable from lab to lab.
• Clear, one piece acrylic top with molded base section
• Double layered, closed cellular neoprene gaskets for airtight seal between top and base
• Adjustable SS clamp compensate for wear
• 8 in. O.D. ports with ambidextrous Hypalon® gloves with SS O-rings
• Transparent transfer chamber (12 L × 11 in. I.D.) with adjustable vacuum gauge
• Gas-drying train with DesiSphere desiccating canisters, self-sealing quick-disconnects enable change of drying train without disturbing internal atmosphere
• Two vacuum/pressure pumps - one for transfer chamber, one for drying train to speed purging
• Fluorescent light system with illuminated controls
Legal Information
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?