• USA Home
  • Z270245 - Programmable daily/weekly time switch
EMAIL THIS PAGE TO A FRIEND
Z270245 Aldrich
Programmable daily/weekly time switch
AC/DC input 120 V, 15 A
Purchase
Properties
Related Categories Extech, Labware, Manufacturer Browser More...
mfr. no.   Extech Instruments 375475
availability   not available in EU
Description
General description
• Program up to 6 ON and 6 OFF switchings
• LCD displays day, time, and program function
• Easy to install and operate
• Single 3-prong plug-in oulet
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?