EMAIL THIS PAGE TO A FRIEND
Z270334 Aldrich
Bunsen burner
Natural gas: Meker
Synonym: bunsen burner gas, bunsen burner lab, bunsen burners
Purchase
Properties
Related Categories Burners, Heating, Labware More...
Description
General description
For Natural Gas. For high temperature work. Nickel plated brass tube with rotatable non-removable air regulator. Perforated metal burner head 25 mm diameter produces an evenly aerated flame of uniform intensity. On heavy stable cast iron base, baked hammertone finish, with serrated gas inlet tube 9 mm diameter.
Laboratory gas burners for use with natural (methane) or cylinder (propane) gases.
Available in 3 styles:
• General-purpose - for general laboratory applications (solid base)
• Fast-flame - with flame stabilizer to control flame temperature and length (three spoke base)
• Meker - for applications where an extremely hot flame is required (only for bottled gas, perforated metal mesh burner)
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?