• USA Home
  • Z270741 - Whatman® Autovials syringeless filters

EMAIL THIS PAGE TO A FRIEND
Z270741 Aldrich

Whatman® Autovials syringeless filters

autovial 12 syringeless filter, glass prefilter, PVDF, 0.2 μm, 50/pk

Synonym: filtration units

Purchase

Properties

Related Categories Analytical/Chromatography, Filtration Supplies, HPLC Sample Preparation, Labware, Manufacturer Browser,
material   PVDF membrane
  polypropylene housing
feature   autoclavable (121°C for 20 minutes)
  disposable
  Outlet connection: male slip luer
packaging   pack of 50
mfr. no.   Whatman Article No., 28421431 (US reference)
  Whatman, AV125EAQU
capacity   12 mL
filter area   3.0 cm2
pore size   0.2 μm

Description

Application

Autovial membranes are available for various applications:
• Cellulose Acetate (CA): low nonspecific protein binding and high loading capacity membrane for biological solutions
• Glass microfiber (GMF): depth filter for samples in aqueous or organic solutions
• Nylon (NYL): for aqueous and organic samples within a pH range between of 3 to 10
• Polyethersulfone (PES): low non-specific protein binding membrane for samples in aqueous solutions
• Polypropylene (PP): Hydrophobic membrane. Resistant to a wide range of organic solvents
• Polytetrafluorethylene (PTFE): PTFE membrane for samples with > 50% organic solvent
• Polyvinylidene Fluoride (PVDF): low nonspecific protein binding membrane for samples in aqueous solutions and/or organic solvents

Features and Benefits

Features and benefits:
• Single unit convenience saves time. No assembly required - easier to load
• Choice of filter media. Compatible with a wide range of sample types
• Ideal for hazardous samples. Self contained device eliminates risk of filter pop-off
• Built-in air purge maximizes sample recovery
• Sterile option available to maintain sample integrity
• Unique prefilter design for difficult-to-filter samples (no prefilter in Autovial 5 and in selected Autovial 12)
• Choice of glass fiber or polypropylene prefilters

General description

Whatman® Autovial Syringeless Filters

Autovial syringeless filters are preassembled filtration devices for removing particulates from samples. They replace syringe-coupled filtration devices with single, disposable units.

Autovial devices are comprised of two parts: a graduated filter barrel and a plunger. The proven design features an integral filter, built-in air purge and a support stand that protects the recessed slip-luer tip. They are available in a 5 mL and 12 mL volume capacity.

The Autovial filter is selected according to membrane compatibility with the sample. In practice, the sample is poured into the 5 mL or 12 mL capacity filter barrel. The plunger is inserted into the barrel until the bottom is securely in place; there is a gap of air between the sample and plunger. Then, the tip of the Autovial is placed into the mouth of an autosampler vial or container and the plunger compressed. Filtration begins immediately and, as the plunger is compressed until it reaches the bottom, the membrane is purged with air for maximum sample recovery. For direct instrument injection, a needle is placed on the Autovial slip-luer outlet.

Other Notes

Global SIAL customers (excluding Japan) can now order any product from the GE Healthcare Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.

The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.

This product has been replaced by WHAAV125EAQU-ALDRICH | Whatman Autovials syringeless filters, autovial 12 syringeless filter, glass prefilter, PVDF, 0.2 μm, 50/pk

Legal Information

Autovials is a trademark of Whatman International Ltd.

Whatman is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

WHAAV125EAQU Whatman® Autovials syringeless filters, autovial 12 syringeless filter, glass prefilter, PVDF, 0.2 μm, 50/pk

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?