• USA Home
  • Z270881 - Whatman® cellulose chromatography papers
EMAIL THIS PAGE TO A FRIEND
Z270881 Aldrich
Whatman® cellulose chromatography papers
3MM Chr roll, 1.5 in x 300 ft, 1/pk
Purchase
Properties
Related Categories Labware, Manufacturer Browser, Whatman, Whatman 3MM papers, Whatman Blotting / Electrophoresis Supplies,
material   cellulose
packaging   pkg of 1 roll
W × L   1.5 in. × 300 ft
Description
General description
Grade 1 Chr
The world standard chromatography paper. A smooth surface, 0.18 mm thick with a linear flow rate (water) of 130 mm/30 min. Good resolution for general analytical separations.
Grade 2 Chr
Thickness 0.18 mm. Flow rate 115 mm/30 min. Slower than 1 Chr for higher resolution applications. Smooth surface. Particularly recommended for optical or radiometric scanning.
Grade 3MM Chr
Though widely used as a blotting paper, 3MM Chr is also used both in electrophoresis and for general chemistry. A medium thickness paper (0.34 mm) used extensively for general chromatography and electrophoresis. Flow rate is 130 mm/30 min.
Grade 4 Chr
Thickness 0.21 mm. Flow rate 180 mm/30 min. Fastest of the thin papers. Recommended for routine and/or repetitive chromatography when loadings are relatively low. Smooth surface. Very suitable where speed is important and very high resolution is not required.
Grade DE 81
DEAE cellulose paper. Weakly basic anion exchanger. Thin paper with medium flow rates.
Grade 2668 Chr
Thickness 0.9 mm. Flow rate 155 mm/10 min. For separation of relatively large molecules by electrophoresis.
Legal Information
Whatman is a registered trademark of GE Healthcare
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?