• USA Home
  • Z271039 - Whatman® high performance cellulose extraction thimbles
EMAIL THIS PAGE TO A FRIEND
Z271039 Aldrich
Whatman® high performance cellulose extraction thimbles
I.D. × H 25 mm × 100 mm
Synonym: Whatman filter, cellulose filter, paper filter
Purchase
Properties
Related Categories Extraction Thimbles, Extractors, Labware, Manufacturer Browser, Physical Properties Testing,
packaging   pkg of 25 ea
mfr. no.   Whatman Article No., 28418966 (US reference)
  Whatman, 2800-250
I.D. × H   25 mm × 100 mm
Description
General description
Cellulose extraction thimbles are produced from high-quality alpha cellulose cotton linter and have excellent mechanical strength and retention. All thimbles are single thickness having a wall thickness of approximately 1 mm, except for Z565636 which is a double wall thickness of approximately 2 mm; good for applications where higher retention and increased wet or dry strength, or rigidity are required.
Other Notes
Global SIAL customers (excluding Japan) can now order any product from the GE Healthcare Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
This product has been replaced by WHA2800250-ALDRICH | Whatman high performance cellulose extraction thimbles, I.D. × H 25 mm × 100 mm
Legal Information
Whatman is a registered trademark of GE Healthcare
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
WHA2800250 Whatman® high performance cellulose extraction thimbles, I.D. × H 25 mm × 100 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?