• USA Home
  • Z271136 - Whatman® high-purity glass microfiber extraction thimbles

EMAIL THIS PAGE TO A FRIEND
Z271136 Aldrich

Whatman® high-purity glass microfiber extraction thimbles

glass thimbles, HP, 19 x 90 mm, 25/pk

Synonym: extraction thimbles, glass fiber filters

Purchase

Properties

Related Categories Extraction Thimbles, Extractors, Labware, Manufacturer Browser, Physical Properties Testing,
material   borosilicate glass
packaging   pack of 25
mfr. no.   Whatman Article No., 28419025 (US reference)
  Whatman, 2814-199
I.D. × H   19 mm × 90 mm

Description

General description

Whatman® High-Purity Glass Microfiber Extraction Thimbles

High-purity glass microfiber thimbles are available for specialized applications. The thimbles are completely free of binders or additives and can be used at temperatures up to 500°C or when using solvents incompatible with cellulose thimbles. These thimbles are also used in pollution monitoring techniques (0.8 μm nominal particle retention). Typical thickness 1.7 mm.

Features and benefits:
• Available in a range of sizes and wall thicknesses to suit your application
• Designed to fit most commercially available Soxhlet extractors
• No binders are added

Applications:
• Smoke stack gas monitoring
• Soxhlet extraction
• Analyzing pesticide residues
• Determining oil/fat content of foods (e.g. French fries)
• Analysis of oil and grease in solid wastes

Other Notes

Global SIAL customers (excluding Japan) can now order any product from the GE Healthcare Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.

The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.

This product has been replaced by WHA2814199-ALDRICH | Whatman high-purity glass microfiber extraction thimbles, glass thimbles, HP, 19 x 90 mm, 25/pk

Legal Information

Whatman is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

WHA2814199 Whatman® high-purity glass microfiber extraction thimbles, glass thimbles, HP, 19 x 90 mm, 25/pk

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?