• USA Home
  • Z271454 - Oxford Chemistry Primers
EMAIL THIS PAGE TO A FRIEND
Z271454 Aldrich
Oxford Chemistry Primers
No. 30: Process Development: Fine Chemicals From Grams to Kilograms
  •  ISBN-10 0198558244
  •  ISBN-13 978-019855824-8
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   S. Lee, Oxford University Press, 1995, 96 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
Process development bridges the gap between the laboratory synthesis of an organic compound and its industrial manufacture on a large scale. This concise and readable text uses real examples and case histories from ICI/Zeneca Pharmaceuticals to show the problems which may be encountered in scaling up chemical synthesis, and the ways these problems may be overcome. It shows how it is possible to synthesize multi-kilogram quantities of a new organic compound which has been made in the laboratory only in the milligram scale. A wide range of aliphatic, aromatic, and heterocyclic compounds is covered. This book will be invaluable both to the research chemist involved in the development phase of novel compounds, and the advanced undergraduate interested in the fine chemicals industry.
Table of Contents
Introduction
1. Scale up issues
2. Environmental and regulatory issyes
3. Fun with cephalosporins
4. Thromboxane antagonists
5. Homochiral thrombozane antagonists
6. H2 blockers
7. Development of a manufacturing route for cloprostenol sodium
8. Synthesis of a potentially anti-obesity agent
9. Reagents for large scale operation
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?