• USA Home
  • Z271470 - Oxford Chemistry Primers
EMAIL THIS PAGE TO A FRIEND
Z271470 Aldrich
Oxford Chemistry Primers
No. 32: Nuclear Magnetic Resonance
  •  ISBN-10 0198556829
  •  ISBN-13 978-019855682-4
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   P. Hore, Oxford University Press, 1995, 96 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
Nuclear magnetic resonance spectroscopy is an enormously powerful and versatile physical method for investigating the structure and dynamics of molecules. This text provides a clear, concise introduction to the physical principles of NMR, and the interactions that determine the appearance of NMR spectra. It describes and explains how nuclear spins interact with a magnetic field (the chemical shift) and with each other (spin-spin coupling); how NMR spectra are affected by chemical equilibria (exchange) and molecular motion (relaxation); and concludes with an outline of the workings of some simple one- and two-dimensional Fourier transform NMR experiments. The ways in which NMR may be used to study the structures, motions and reactions of molecules are illustrated and discussed. Only essential mathematics and theory are presented. The emphasis throughout is on understanding the basic principles
Table of Contents
1. Introduction
2. Chemical shifts
3. Spin-spin coupling
4. Chemical exchange
5. Spin relaxation
6. Experimental methods
Bibliography
Index
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?