• USA Home
  • Z271500 - Quality in the Analytical Chemistry Laboratory
EMAIL THIS PAGE TO A FRIEND
Z271500 Aldrich
Quality in the Analytical Chemistry Laboratory
  •  ISBN-10 0471954705
  •  ISBN-13 978-047195470-5
Purchase
Description
General description
Introduces the whole concept of quality assurance and implementation. Discusses how sampling, method selection, equipment, and taking measurements affects the quality of analytical data.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?