• USA Home
  • Z271748 - Logic Of Chemical Synthesis 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z271748 Aldrich
Logic Of Chemical Synthesis 2nd ed.
  •  ISBN-10 0-471115940
  •  ISBN-13 978-0-47111594-6
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   E.J. Corey, John Wiley & Sons, Ltd, 1995, soft cover
description   2nd edition
mfr. no.   Wiley
Description
General description
This book provides a detailed description of the general principles and strategies for solving chemical synthesis problems of complex organic molecules. It is the only book to deal systematically with the fundamental principles of synthetic problem solving.
Table of Contents
GENERAL APPROACHES TO THE ANALYSIS OF COMPLEX SYNTHETIC PROBLEMS.
The Basis for Retrosynthetic Analysis.
Transform-Based Strategies.
Structure-Based and Topological Strategies.
Stereochemical Strategies.
Functional Group-Based and Other Strategies.
Concurrent Use of Several Strategies.
SPECIFIC PATHWAYS FOR THE SYNTHESIS OF COMPLEX MOLECULES.
Macrocyclic Structures.
Heterocyclic Structures.
Sesquiterpenoids.
Polycyclic Isoprenoids.
Prostanoids.
Leukotrienes and Other Bioactive Polyenes.
GUIDE TO THE ORIGINAL LITERATURE OF MULTISTEP SYNTHESIS.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?