• USA Home
  • Z272310 - Aroma Biotechnology
EMAIL THIS PAGE TO A FRIEND
Z272310 Aldrich
Aroma Biotechnology
  •  ISBN-10 3540586067
  •  ISBN-13 978-354058606-7
Purchase
Properties
Related Categories Books and Software, Flavors and Fragrances Books, Labware More...
publication info   R. Berger, Springer Verlag, 1995, 240 pp., hard cover
mfr. no.   Springer
Description
General description
Aroma biotechnology opens access to natural volatile flavors. Due to the attribute of "naturalness" these aromas are the most valuable ingredients for foods, cosmetics and related products. The author describes biocatalyzed reactions and biogenetic routes leading to aroma and flavor compounds and he depicts in detail their utilisation in industrial scale processes. All the important topics, including de-novo-synthesis, genetic engineering, in vitro rDNA methods, laboratory requirements and techniques, upscaling, product recovery, profitability, and legal aspects are covered. R.G. Berger contributes his own experimental experience to all except one chapter and leads the reader into state-of-the-art aroma biotechnology.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?