• USA Home
  • Z272426 - Preparative TLC plates

EMAIL THIS PAGE TO A FRIEND
Z272426 Aldrich

Preparative TLC plates

C18-silica gel matrix

  •  eCl@ss 32110504

Purchase

Properties

Related Categories Analytical/Chromatography, Chromatography, Chromatography, TLC Plates, LPLC/TLC/Paper Chromatography, Labware,
material   C18-silica gel matrix
  glass support
feature   binder Inorganic
  fluorescent indicator
packaging   pkg of 25 ea
mfr. no.   Analtech, 52013
application(s)   thin layer chromatography (TLC): suitable
L × W   20 cm × 20 cm
layer thickness   1,000 μm

Description

General description

Used for analytical separations of larger quantities of materials. Materials to be separated are often applied as long streaks, developed, and recovered by scraping the adsorbent from the plate and eluting with a strong solvent.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?