• USA Home
  • Z272655 - SET-TEMP® 316 SS thermocouple probe
EMAIL THIS PAGE TO A FRIEND
Z272655 Aldrich
SET-TEMP® 316 SS thermocouple probe
probe type, K, 316 stainless steel sheath
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Controllers, Barnstead Thermolyne Heating Equipment, Heating, Heating Controllers,
material   316 stainless steel sheath
feature   probe type K
mfr. no.   Barnstead International 8303
Description
General description
Sheaths are 6.4 mm (1/4 in.) diameter × 305 mm (12 in.) L to fit common thermometer adapters. Probes have 183 cm (6 ft) lead wires.
Legal Information
SET-TEMP is a registered trademark of Laboratory Devices, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?