• USA Home
  • Z272698 - SET-TEMP® 316 SS thermocouple probe
EMAIL THIS PAGE TO A FRIEND
Z272698 Aldrich
SET-TEMP® 316 SS thermocouple probe
probe type, K, PTFE-coated sheath
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Controllers, Barnstead Thermolyne Heating Equipment, Heating, Heating Controllers,
material   PTFE-coated sheath
feature   probe type K
mfr. no.   Barnstead International 8306
Description
General description
Sheaths are 6.4 mm (1/4 in.) diameter × 305 mm (12 in.) L to fit common thermometer adapters. Probes have 183 cm (6 ft) lead wires.
Legal Information
SET-TEMP is a registered trademark of Laboratory Devices, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?