• USA Home
  • Z272736 - Nalgene® PFA beaker
EMAIL THIS PAGE TO A FRIEND
Z272736 Aldrich
Nalgene® PFA beaker
capacity 30 mL
Purchase
Properties
Related Categories Beakers, Labware, Manufacturer Browser, Nalgene, Nalgene Beakers,
material   PFA
mfr. no.   Nalgene
capacity   30 mL
Description
General description
PFA unaffected by most chemicals. Withstands a temperature range from -270 to 250°C.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?