• USA Home
  • Z272949 - FT-IR Spectral Libraries
EMAIL THIS PAGE TO A FRIEND
Z272949 Aldrich
FT-IR Spectral Libraries
Nicolet®/Aldrich Condensed Phase Library, edition 2
Purchase
Properties
Related Categories Computer Software, Labware, Spectral Libraries More...
Description
General description
Libraries may be used with OMNIC®, GRAMS®/32, or Spectral I.D.® (Z407488) and are supplied on CD-ROM. Libraries are in deresolved format and contain spectra at 16 cm-1 resolution by 8-bit ordinate precision.
• Contain spectral data from 4,000 to 450 cm-1
• Best used when searching compounds with significant spectral differences
• Generally not suitable when using spectral subtraction or other precision spectral identification techniques
The expanded Aldrich FT-IR Condensed Phase Library, Edition 2 now contains nearly 18,500 pure compounds – the most comprehensive FT-IR spectral reference collection available. The collection contains the most common chemicals found in the Aldrich Handbook, many of which are pure organic compounds, and includes the New Second edition of the Aldrich Library of FT-IR Spectra (3-volume set). All spectra were acquired and examined by Aldrich and then re-examined and processed by Nicolet. This is the largest collection of spectra collected entirely by FT-IR instrumentation.
Other Notes
Other libraries are available –please inquire.
• Alcohols and phenols
• Aldehydes and ketones
• Carboxylic acids, esters, lactones, and anhydrides
• Polymers
• Solvents
• More
Legal Information
GRAMS is a registered trademark of Thermo Electron Corp.
Nicolet is a registered trademark of Thermo Electron Corp.
OMNIC is a registered trademark of Thermo Electron Corp.
Spectral ID is a registered trademark of Galactic Industries Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?