• USA Home
  • Z273015 - FT-IR Spectral Libraries
EMAIL THIS PAGE TO A FRIEND
Z273015 Aldrich
FT-IR Spectral Libraries
Nicolet® Polymers, Polymer Additives & Plasticizers Condensed Phase Library
Purchase
Properties
Related Categories Computer Software, Labware, Spectral Libraries More...
mfr. no.   Nicolet
Description
General description
Includes more than 1,750 spectra of polymers, plastics and additives for polymers, rubbers, cosmetics, adhesives, sealants, and plastics. Sequential index includes: manufacturer, chemical name, CAS number, and more. Collected by Chemir/Polytech Laboratories and Dr. John Kokosa.
Libraries may be used with OMNIC®, GRAMS®/32, or Spectral I.D.® (Z407488) and are supplied on CD-ROM. Libraries are in deresolved format and contain spectra at 16 cm-1 resolution by 8-bit ordinate precision.
• Contain spectral data from 4,000 to 450 cm-1
• Best used when searching compounds with significant spectral differences
• Generally not suitable when using spectral subtraction or other precision spectral identification techniques
Other Notes
Other libraries are available –please inquire.
• Alcohols and phenols
• Aldehydes and ketones
• Carboxylic acids, esters, lactones, and anhydrides
• Polymers
• Solvents
• More
Legal Information
GRAMS is a registered trademark of Thermo Electron Corp.
Nicolet is a registered trademark of Thermo Electron Corp.
OMNIC is a registered trademark of Thermo Electron Corp.
Spectral ID is a registered trademark of Galactic Industries Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?