• USA Home
  • Z273023 - FT-IR Spectral Libraries
EMAIL THIS PAGE TO A FRIEND
Z273023 Aldrich
FT-IR Spectral Libraries
Nicolet® Food Additives Condensed Phase Library
Purchase
Description
General description
Libraries may be used with OMNIC®, GRAMS®/32, or Spectral I.D.® (Z407488) and are supplied on CD-ROM. Libraries are in deresolved format and contain spectra at 16 cm-1 resolution by 8-bit ordinate precision.
• Contain spectral data from 4,000 to 450 cm-1
• Best used when searching compounds with significant spectral differences
• Generally not suitable when using spectral subtraction or other precision spectral identification techniques
Other Notes
Other libraries are available –please inquire.
• Alcohols and phenols
• Aldehydes and ketones
• Carboxylic acids, esters, lactones, and anhydrides
• Polymers
• Solvents
• More
Legal Information
GRAMS is a registered trademark of Thermo Electron Corp.
Nicolet is a registered trademark of Thermo Electron Corp.
OMNIC is a registered trademark of Thermo Electron Corp.
Spectral ID is a registered trademark of Galactic Industries Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?