• USA Home
  • Z273120 - High-capacity variable take-off splitter head
EMAIL THIS PAGE TO A FRIEND
Z273120 Aldrich
High-capacity variable take-off splitter head
Purchase
Description
General description
One-piece jacketed design features a Torion adjusting valve, heavy-duty condenser, and jacketed drip-tip joint. Designed to handle higher volume reflux and higher temperature liquids. The column and receiver joints 24/40 with a 10/30 thermometer joint. Two 4 mm glass stopcocks, PTFE 0-3 mm metering valve.
Legal Information
Torion is a trademark of Bibby Sterlin Ltd.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?