• USA Home
  • Z273686 - Free Radicals In Organic Chemistry

EMAIL THIS PAGE TO A FRIEND
Z273686 Aldrich

Free Radicals In Organic Chemistry

  •  ISBN-10 0471954969

  •  ISBN-13 978-047195496-5

Purchase

Properties

Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   J. Fossey, John Wiley & Sons, Ltd, 1995, 322 pp., soft cover
mfr. no.   Wiley

Description

General description

Illustrious researchers offer an excellent introduction to the structure, formation and reactivity of free radicals. Contains a complete overview of the numerous and varied applications of free radicals within organic chemistry with special attention to reaction mechanisms and novel responses. Includes detailed accounts along with actual examples of the most recent developments in the field.

Table of Contents

PARTIAL TABLE OF CONTENTS:
GENERAL CONCEPTS AND BASIC PRINCIPLES.
Detection and Observation of Free Radicals.
Radical Structure.
Radical Stability.
Reactivity of Free Radicals.
Radical Kinetics.
REACTIONS: CLASSIFICATIONS AND MECHANISMS.
Production of Free Radicals.
Radical-Radical Reactions.
Substitution Reactions.
Addition and Fragmentation Reactions.
Reactions of Charged Radicals.
Free Radicals in Biochemistry.
APPLICATIONS IN SYNTHESIS.
Functionalization of C-H Bond.
Transformation of Functional Groups.
Cyclization.
Aromatic Substitution.
Coupling Reactions.
Free Radical Reactions in Industry.
REFERENCES AND TABLES.
References.
Index.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?