• USA Home
  • Z274879 - Maintenance kit for KNF Laboport® solid PTFE vacuum pump
EMAIL THIS PAGE TO A FRIEND
Z274879 Aldrich
Maintenance kit for KNF Laboport® solid PTFE vacuum pump
Maintenance kit for 20 Lpm pump
Purchase
Description
General description
Includes replacement PTFE diaphragm, head seals and two Kalrez valves.
Kits include replacement PTFE diaphragm, head seals, and two Kalrez® valves. Note: Two kits are required when servicing both heads of vacuum pump.
Other Notes
Two kits are required when servicing both heads of vacuum pump.
Legal Information
Kalrez is a registered trademark of E. I. du Pont de Nemours and Company
Laboport is a registered trademark of KNF Neuberger GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?