• USA Home
  • Z275425 - Modified Ubbelohde viscometer tube
EMAIL THIS PAGE TO A FRIEND
Z275425 Aldrich
Modified Ubbelohde viscometer tube
capillary section size 0.7 mm
Purchase
Properties
Related Categories Ace, Ace Viscometers, Labware, Manufacturer Browser, Tubes,
mfr. no.   Ace Glass 798710
capillary section size   0.7 mm
Description
General description
Directions given for relative, inherent, and intrinsic viscosities. Used in ASTM Method D1601 for ethylene polymers having densities from 0.913 to 0.970g/cc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?