Synonym: cuvette quartz, quartz cell, quartz cuvette
| Related Categories | Absorption Cells, Analytical Reagents, Analytical/Chromatography, Cuvettes, Labware, |
| material | glass |
| limit | 334-2,500 nm spectral range |
| H × W × D | 45 mm × 12.5 mm × 12.5 mm |
• Solid and open-top cap styles
• Protect air-sensitive samples
• Prevent evaporation and contamination
• Avoid potential spills and exposure to chemicals
Available as a solid-top screw-cap with silicon rubber liner and an open-top screw-cap with silicon rubber septum. Has 2 clear windows and 2 frosted windows. Standard 3.5 mL rectangular cells. Light path: 10 mm. Fits all standard spectrophotometers.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?