| Related Categories | Absorption Cells, Analytical Reagents, Analytical/Chromatography, Cuvettes, Glass spectrophotometer cuvettes, |
| material | optical glass |
| limit | 334-2,500 nm spectral range |
| chamber volume | 700 μL |
| pathlength | 10 mm |
Rectangular cells for work in the visible range with windows of optical glass. With PTFE cover.