Z276952 Aldrich

Raman quartz cuvette

chamber volume 3.5 mL, cover screw top (open)

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Cuvettes, Labware, Manufacturer Browser,
material   cover screw top (open)
  quartz
limit   170-2700 nm spectral range
H × W × D   45 mm × 12.5 mm × 12.5 mm
chamber volume   3.5 mL

Description

General description

Fluorescence-free rectangular cells are polished on all four sides and bottom. Solid screw-caps have a PTFE liner, open screw-caps have a PTFE-lined silicone septum.

Price and Availability

Customers Also Viewed

Raman quartz cuvette

volume 1.4 mL, PTFE cover

Raman quartz cuvette

volume 3.5 mL, PTFE cover

Raman quartz cuvette

volume 3.5 mL, cover screw top (solid)

Screw-cap cuvettes

Spectrosil quartz, Solid-top cap, limit 170-2,700 nm spectral range

UV quartz cuvette

Semi-micro rectangular, volume 1.4 mL, PTFE cover

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?