• USA Home
  • Z277592 - Compilation of EPA’s Sampling and Analysis Methods, 2nd. ed.
EMAIL THIS PAGE TO A FRIEND
Z277592 Aldrich
Compilation of EPA’s Sampling and Analysis Methods, 2nd. ed.
  •  ISBN-10 1566701708
  •  ISBN-13 978-156670170-9
Purchase
Properties
publication info   L. Keith, CRC Press, 1997, 1728 pp., hard cover
mfr. no.   CRC Press
Description
General description
The first edition contained approximately 600 analyte/method summaries. This edition contains twice as many new EPA-approved methods for testing and analyzing industrial chemicals, pesticides, herbicides, dioxins, and PCBs and is a printed version of the EPA′s Sampling and Analysis Methods Database. Each analyte/method summary contains all of the information required to stand alone as a reference. Thus, in addition to a brief summary of each method, descriptions include required instrumentation, interferences, sampling containers, preservation techniques, maximum holding times, detection levels, accuracy, precision, quality control requirements, EPA reference, and, when available, EPA contacts with phone numbers. Each summarized report is a "stand-alone" document.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?