• USA Home
  • Z277762 - HGS Polyhedron molecular model
EMAIL THIS PAGE TO A FRIEND
Z277762 Aldrich
HGS Polyhedron molecular model
student set, student crystal structure-H
Purchase
Properties
Related Categories HGS, HGS models, Labware, Manufacturer Browser, Molecular Models,
description   scale: 1 Å = free
feature   no. of atoms 84
  no. of bonds 190
mfr. no.   HGS 1004
Description
General description
The use of 14, 20, and 26-sided polyhedrons as atomic nuclei makes these models unique. The various bond angles and distances needed for the models can be precisely represented, and the symmetrical characteristics of the structures can be observed.
• Color-coded polyhedrons to indicate different elements
• Flexible polyacetal connecting bonds permit easy assembly of complicated molecules and models of strained compounds and distorted crystals
• Å scale provided with sets allows measurement of bond distances
• Bond puller included for easy disassemble
• Two types of orbital plates for p atomic orbitals and π molecular orbitals (some sets only)
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?