• USA Home
  • Z277797 - HGS Researcher model set
EMAIL THIS PAGE TO A FRIEND
Z277797 Aldrich
HGS Researcher model set
Crystal structure-E
Purchase
Properties
Related Categories HGS, HGS models, Labware, Manufacturer Browser, Molecular Models,
feature   no. of atoms 292
  no. of bonds 578
mfr. no.   HGS 1007
Description
General description
Contain a variety of polyhedrons and bonds to meet the broad demand of chemical research. Scale: 1Å = 2.5 cm.
Researcher organic set is suited for building fundamental organic compounds, polymers, steroids, and some simple complex salts.
Researcher inorganic set is suited for building complicated tetra-, hexa-, and octa-coordinated complex compounds and chelate compounds. The fundamental structure of ionic crystals such as rock salt, cesium chloride, etc. can be constructed without supplementary parts.
Crystal structure set permits assembly of nearly all the fundamental crystal structures in inorganic chemistry and crystallography, as well as inorganic giant molecules.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?