• USA Home
  • Z277800 - HGS Polyhedron molecular model
EMAIL THIS PAGE TO A FRIEND
Z277800 Aldrich
HGS Polyhedron molecular model
lecturer set, organic chemistry-G
Purchase
Properties
Related Categories HGS, HGS models, Labware, Manufacturer Browser, Molecular Models,
description   scale: 1 Å = 7.5 cm
feature   no. of atoms 84
  no. of bonds 108
  no. of orbital plates 18
mfr. no.   HGS 1008
Description
General description
The giant size of these models, makes them well suited for lectures and demonstrations in a large room. Strong, lightweight aluminum bonds maintain the structure of large molecules. Set contains two types of orbital plates designed for p atomic and π molecular orbitals.
The use of 14, 20, and 26-sided polyhedrons as atomic nuclei makes these models unique. The various bond angles and distances needed for the models can be precisely represented, and the symmetrical characteristics of the structures can be observed.
• Color-coded polyhedrons to indicate different elements
• Flexible polyacetal connecting bonds permit easy assembly of complicated molecules and models of strained compounds and distorted crystals
• Å scale provided with sets allows measurement of bond distances
• Bond puller included for easy disassemble
• Two types of orbital plates for p atomic orbitals and π molecular orbitals (some sets only)
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?