• USA Home
  • Z277835 - HGS Stereochemistry Molecular Model Set
EMAIL THIS PAGE TO A FRIEND
Z277835 Aldrich
HGS Stereochemistry Molecular Model Set
HGS 4000
Purchase
Description
General description
It is featherweight, slim, and flexible, and yet quite durable-ideal not only for research chemists but also for learning students as well. A new scale, 1 Å = 2 cm, offers easy handing and assures the precise estimation of internulear distances. Conformational changes can be attained easily and smoothly. Yet any specific conformer can be fixed without any special aid. The flexible nature of bonding allow ones to construct four-and five-membered rings using a usual unit. The model is suitable in the fields of organic chemistry, biochemistry, and inorganic chemistry both for educational and research purposes.
Scale: The newly adopted scale, 1 Å = 2 cm, makes the model handy and varsatile.
Materials and Parts:
Standard set 4000: Made of superb polyacetal resin. More than ten basic units, all color-coded, are available. Special units are also supplied, which include a six-coordinated atom unit and a hydrogen-bond unit.
Advanced set 7000: same as set 4000 but contains the following additional parts: cyclopropane, epoxide, aziridine, cyclobutene, benzene, naphthalene, porphyrin, and two types of octahedral metal (one for conventional use and the other for porphyrin metalation) Supplied in a plastic case.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?