• USA Home
  • Z277851 - HGS Biochemistry molecular models
EMAIL THIS PAGE TO A FRIEND
Z277851 Aldrich
HGS Biochemistry molecular models
Bio-organic set
Purchase
Description
General description
Suitable for constructing α -helix and β -sheet of twenty peptide backbones and two nucleotide pairs, adenine-thymine and guanine-cytosine. 264 pieces.
These miniature-size models are low in cost, easy to assemble, and portable. Useful for teaching organic chemistry and biochemistry, and for laboratory work. Important biochemical groups are assembled into units, such as peptide bonding units, ribose, and deoxyribose ring units, for easy addition to other parts and units. Color-coded parts are made of flexible polyacetal resin. Can be used interchangeably with HGS Space-filling molecular models. Scale: 1 Å = 1.0 cm.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?