• USA Home
  • Z278912 - Fruit Flavors: Biogenesis, Characterization, and Authentication
EMAIL THIS PAGE TO A FRIEND
Z278912 Aldrich
Fruit Flavors: Biogenesis, Characterization, and Authentication
  •  ISBN-10 084123227X
  •  ISBN-13 978-084123227-3
Purchase
Properties
Related Categories Books and Software, Flavors and Fragrances Books, Labware More...
publication info   R. Rouseff and M. Leahy, American Chemical Society, 1995, 308 pp., hard cover
mfr. no.   American Chemical Society
Description
General description
Examines new analytical techniques to determine authenticity of "natural" fruit flavors, aromas, and essential oils. Describes the biochemical pathways as well as bioconversion of fruit flavors. Reviews the conversion of flavorless precursors to intensely flavored key aroma components in tomatoes, apricots, and apples and describes the effect of packaging and storage on flavor of orange juice, apples, pears, and berries. Describes analytical and sensory characteristics of orange juice, cupuac, carambola, and kiwi fruit flavors and correlates analytical and sensory techniques. Explores the effect that environmental factors, maturity, and variety can have on the quality of fruit products.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?