• USA Home
  • Z281409 - Uvex® Stealth safety goggles
EMAIL THIS PAGE TO A FRIEND
Z281409 Aldrich
Uvex® Stealth safety goggles
gray frame
Purchase
Properties
Related Categories Eye Protection Equipment, Goggles, Labware, Safety Equipment
availability   not available in EU
  not available in EU
color   gray frame
Description
General description
• Wide curve antifog lenses for unrestricted field of view
• Unique ventilation system keeps out splashes, particulates, and vapors
• Internal silicone rubber cushion provides full-day comfort
Clear PC lens with 4-C coating (antifog, antistatic, antiscratch, and UV). UV protection blocks 99.9% of harmful UV rays. Meets ANSI Z87.1-1989 and exceeds ANSI Z80.3 requirements.
Legal Information
Stealth is a trademark of Bacou-Dalloz Eye & Face Protection, Inc.
Uvex is a registered trademark of Uvex Winter Optik GmbH Ltd.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?