EMAIL THIS PAGE TO A FRIEND
Z281581 Aldrich
Radiation meter
battery power 9 V (battery included)
Purchase
Description
General description
Sensitive enough to detect minuscule amounts of β, γ or x-rays. Excellent for scanning samples, work areas, and clothing for traces of radioactivity. Can be used as a personal monitor in areas of potential exposure. Uses a Geiger-Mueller tube detector with a range of 0.1 to 10.0 mR/hr or 0.1 to 100 μSv/h. Audible clicking upon detection of radiation; monitor will beep at 20.0 mR/hr.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?