• USA Home
  • Z282219 - QuickTRAP vacuum protection system

EMAIL THIS PAGE TO A FRIEND
Z282219 Aldrich

QuickTRAP vacuum protection system

Purchase

Description

General description

QUICKTRAP systems are designed to trap contaminants and biological wastes that have been aspirated in a vacuum system. The 2L QUICKTRAP features a 3/8 in. thick acrylic cabinet for protection from beta-emitting isotopes.
Safety Features:
• All plastic construction eliminates implosion hazard
• Primary and secondary traps
• Easy to open and empty
• PTFE filter stops overflow
• Protection from beta-emitting isotopes (2 L QUICKTRAP only)
Specifications:
Reservoirs: Heavy-duty Nalgene® PP bottles, chemically resistant and autoclavable. Continuous use at house vacuum (0.68 bar) over 24-hour period at 20 °C. Closures have TPE gaskets.
Filter: Hydrophobic PTFE membrane, 50 mm diam., 0.2 μm, with PP housing
Tubing: Tygon® vacuum tubing with in-line ABS quick-disconnect fittings
Housing: Acrylic cabinet

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

QuickTRAP is a trademark of C.B.S. Scientific Co.

Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?