| Related Categories | Diaphragm Vacuum Pumps, Labware, Pumps, Vacuum Pump Accessories, Vacuum Pumps More... |
| volume | 5 L |
QUICKTRAP systems are designed to trap contaminants and biological wastes that have been aspirated in a vacuum system. The 2L QUICKTRAP features a
Safety Features:
• All plastic construction eliminates implosion hazard
• Primary and secondary traps
• Easy to open and empty
• PTFE filter stops overflow
• Protection from beta-emitting isotopes (2 L QUICKTRAP only)
Specifications:
Reservoirs: Heavy-duty Nalgene® PP bottles, chemically resistant and autoclavable. Continuous use at house vacuum (0.68 bar) over 24-hour period at 20 °C. Closures have TPE gaskets.
Filter: Hydrophobic PTFE membrane, 50 mm diam., 0.2 μm, with PP housing
Tubing: Tygon® vacuum tubing with in-line ABS quick-disconnect fittings
Housing: Acrylic cabinet
supplied with 1 x 1L and 1 x 4Lbottles
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
QuickTRAP is a trademark of C.B.S. Scientific Co.
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.
Customers Also Viewed
volume 2 L
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?